Would you like to log in again?
Chemical Storage Start-Up Kit, Ward's
Supplier: Ward's
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ward's®
WARD470010-858
470010-858EA 42.99 USD InStock
470010-858
Chemical Storage Start-Up Kit, Ward's
Labels
This kit maintain a safe environment by storing chemicals properly.
  • Detailed instructions
  • Chemical storage posters
  • Color-coded system
This kit helps you create a consistent storage classification system. The provided color-coding system allows you to label chemicals for correct use and storage and by compatibility, so you have an effective storage system that’s compliant with industry standards.
For each of the five colors used in this system (green, white, yellow, red, and blue), there are enough label dots included to re-label 350 bottles and sixty-nine 3' shelves.
Certifications: OSHA/WHMIS-Compliant.
Ordering information: This kit includes rolls of tape and label dots, detailed instructions, one large Ward's chemical storage system poster, and four small posters ¹/₂.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty