Would you like to log in again?
Where Has My Little Dog Been? Kit
Supplier: Avantor
WARD470012-640
470012-640EA 458.79 USD InStock
470012-640
Where Has My Little Dog Been? Kit
Educational Classroom Kits and Activities
Use multiple forms of evidence analysis.
  • Test the evidence and determine the culprit
  • Six different activities
Poor puppy Plato was dognapped. Now that he is safely back home, it’s time to test the evidence and determine the culprit of the canine caper. While testing the evidence and completing six different activities, students learn about blood typing, fiber identification, osmosis, polymers, alloys, solubility, and precipitates. Developed by Nancy Fischer, the activity includes materials for 12 setups and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty