Would you like to log in again?
Grafco® Attached Blood Pressure Set
Supplier: GRAHAM-FIELD INC
Grafco®
242
470012-796EA 66.8 USD InStock
470012-796
Grafco® Attached Blood Pressure Set
Educational Materials Biology Educational Materials Physiology Learning Activities
Perfect for a student team.
  • Cotton cuff and latex-free two (2) tube bladder
  • Self-adjusting D-Bar cuff
  • Pressure measurement from 20 mm Hg to 300 mm Hg
  • Chest piece attached to the cuff for consistent placement
An attached stethoscope allows for more consistent readings and placement for students for training purposes.
Delivery information: The set includes a no-pin, aneroid sphygmomanometer, a self-adjusting cuff with gauge holder, and an attached high-sensitivity nurse stethoscope. All items are supplied in a convenient vinyl storage case.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty