Would you like to log in again?
Acrylic Double Concave/Convex Lens
Supplier: United Scientific Supplies
PACV86 PACC86
470012-814EA 17.75 USD InStock
470012-814 470106-854
Acrylic Double Concave/Convex Lens
Educational Materials Physics Educational Materials Optics Learning Activities
Frosted acrylic lenses for optical experiments.
  • Frosted on one side
  • 15 mm thick
Acrylic Double Concave Lens
Made from clear acrylic. One long plane surface is frosted to make the light beam visible. The lens is 86 mm long, 15 mm thick, and 18 mm wide at the center.
Acrylic Double Concave Lens
Made from clear acrylic. One plane surface is frosted. The lens is 89 mm long, 15 mm thick, and 25 mm across the beam axis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty