Would you like to log in again?
Hollow Prism
Supplier: United Scientific Supplies
PHA045
470012-844EA 10.8 USD InStock
470012-844
Hollow Prism
Educational Materials Physics Educational Materials Optics Learning Activities
Perform Several Optics Experiments
  • This unique optics kit comes with a clear hollow prism, funnel, and plug
  • Measures 2 1/2" H x 1 7/8" W x 1 13/16" D
Young scientists can fill it with clear or colored liquid to explore light refraction and the visible light spectrum. A perfect addition to the classroom for demonstrations or science projects.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty