Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ward's® Chemistry Model 1000 Spectrophotometer

Supplier: UNITED PRODUCT & INSTRUMENT SE
S-1000VWR S-1100-103
470017-376EA 27.3 USD InStock
470017-376 470013-168
Ward's® Chemistry Model 1000 Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Precisely designed and ruggedly built, this easy-to-use spectrophotometer with a large digital display is ideal for student use in high schools and colleges.

  • USB Computer Port
  • Wavelength Range: 400–1000 nm with ±3 nm Accuracy
  • Photometric Range: 0–100% T, 0–1.99 A with ±2% Accuracy

It is suitable for general analysis and experiments such as Beer-Lambert, absorption spectrum, chlorophyll, protein (biuret test), and more. The built-in secondary filters reduce stray light and increase precision.

The student-proof positive safety stopper integrated with the wavelength dial prevents damage to the monochromator and grating. New features include a USB computer port.

Ordering information: It comes with a 1/2" round tube holder, 10 mm square cuvette adapter, 12 test tubes, user’s manual, and dust cover.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty