Would you like to log in again?
Friction Apparatus
Supplier: United Scientific Supplies
FRAP01
470013-276EA 51.8 USD InStock
470013-276
Friction Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Versatile Unit Tests Several Factors of Friction
  • Investigate the affect of weight on frictional forces
  • Includes an Activity Guide
This set includes a friction block and a friction board, both made of high quality, smoothly finished pine wood. The friction block has a hook for a spring scale and two compartments for additional weights. It can be placed face-up or sideways. Students can vary the weight or area of contact to see how they impact the friction force. It also includes three 50 g weights, a 100 g spring scale, and a comprehensive activity manual. The board is 20" x 2" and the block is 4" x 1 1/2" x 1".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty