Would you like to log in again?
Flame Test Station Kit
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470013-550
470013-550EA 62.99 USD InStock
470013-550
Flame Test Station Kit
Educational Classroom Kits and Activities
Setting Up Your Flame Test Lab Just Got Easier
  • Kit contains all necessary materials for lab set up
  • Further experiments can be performed with spectroscopes you provide
This kit has everything you need, conveniently arranged and clearly explained. Simply open up the five included bottles of solutions, Lay out the wood splints, pass out the instructions and you are on your way to a successful lab activity. Cobalt glass is also included to help distinguish between the potassium and the sodium flame test.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty