Would you like to log in again?
Ward's® Chemistry Measurements Using Electrochemical Cells and Electroplating Lab Activity
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470013-610
470013-610EA 109.99 USD InStock
470013-610
Ward's® Chemistry Measurements Using Electrochemical Cells and Electroplating Lab Activity
Educational Classroom Kits and Activities
This lab teaches students about oxidation-reduction reactions through electroplating.
  • Activity includes Electrolyzing Copper Sulfate Solution with an Electrolytic Cell
  • Students Use Empirical Evidence from Experiment to Calculate Amount of Oxidized Copper
  • Required Materials Included
  • Lab Covers a Significant Industrial Process: Electroplating
Electroplating is a valuable industrial process that allows metals to be plated onto other materials using an electrical current. In this activity, an electrolytic cell is used to electrolyze a solution of copper (II) sulfate. A voltage source connects to two copper strips, and the strips are placed into the solution of copper ions. The energy from the voltage source causes a redox reaction to occur within the cell.
At one electrode, copper (II) ions reduce to elemental copper, and at the other electrode, elemental copper oxidize to form copper (II) ions. When students measure the initial and final masses of the two electrodes, they can calculate the amount of copper that oxidized and reduced in the experiment.
Ordering information: This lab includes copper sulfate pentahydrate, copper metal strips, and sulfuric acid solution. Ammeter, an analytical balance, a 150 mL beaker, a battery, a timer, a 100 mL graduated cylinder, isopropyl alcohol, wires, and alligator clips are required but not included.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty