Would you like to log in again?
Ward's® Chemistry Polymer Worms Demonstration
Supplier: Avantor
WARD470013-626
470013-626EA 58.99 USD InStock
470013-626
Ward's® Chemistry Polymer Worms Demonstration
Educational Classroom Kits and Activities
A Fun Real-Life Application for Cross-linking and Polymer Chemistry
  • Two kit options available: teacher demonstration or student activity
  • Required basic labware that you provide.
This activity demonstrates how a simple material extracted from seaweed is used as a common thickening agent for popular candies. Sodium alginate is a long-chain, natural polysaccharide polymer that contains sodium ions. When placed in a solution of calcium chloride, the divalent calcium ions displace the sodium ions and the polymers are quickly cross-linked to form a worm-shaped gel. These gelatinous worms can be safely examined since the polymer is a nontoxic food additive. The demonstration includes enough materials to for 10 repetitions. The activity contains enough materials for 12 student groups to prepare several worms.
Ordering information: Includes Sodium Alginate, Calcium Chloride, Food Coloring, and Syringe. General Glassware/Labware are required but not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty