Would you like to log in again?
Mole In The Bag
Supplier: Avantor
WARD470013-628
470013-628EA 76.99 USD InStock
470013-628
Mole In The Bag
Educational Classroom Kits and Activities
How Big Is A Mole? Why does a mole of copper weigh more than a mole of sulfur?
  • No Additional Equipment Needed
  • Use year after year
Students will answer these inquiry questions and more while introducing the concept of the mole and its importance in chemistry with this visual demonstration. The bags contain several elements and compounds and are available for students to examine, weigh, and explore. Use the mini bags to demonstration fractions of a mole, then bring the bags out again to demonstrate the relationship between moles and molar solutions. This is a great teaching aid for introducing moles, Avogadro's Number, gram molecular weight, and morality.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty