Would you like to log in again?
Demonstration Bimetallic Strip
Supplier: VWR International
PH 0403A
470014-318EA 26.6 USD InStock
470014-318
Demonstration Bimetallic Strip
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Vivid Illustration of Thermoexpansion.
A 12V halogen bulb is mounted below this metallic strip to heat it. Once heated, the strip bends upward and breaks the circuit.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty