Would you like to log in again?
Fundamentals of Physics Pulley System
Supplier: VWR International
RPL08
470014-338EA 94.9 USD InStock
470014-338
Fundamentals of Physics Pulley System
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Discover the Mechanical Advantage of Different Pulley Combinations
  • Multiple pulleys in one comprehensive device, this system allows students to experiment with many variations of string length, force, and more
  • Requires the Fun stand, sold separately
Demonstrate that the length of string pulled increases with the number of pulleys used. Also calculate the work conserved between input and output work when the amount of force put into the system decreases with the more pulleys students add.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty