Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Economy Force Table

Supplier: VWR International
PH0347CN8
470014-356EA 224.35 USD InStock
470014-356
Economy Force Table
Educational Materials Physics Educational Materials Mechanics Learning Activities
The heavy base of the economy force table offers stability for all applications.

  • For all classroom applications
  • Laminated wood tabletop
  • Angle scale marked in 1° increments
  • Four 50 g mass hangers
  • Four table-clamp pulleys

A laminated wood tabletop features an angle scale marked in 1° increments.

Each set of slotted masses has five 20 g, one 10 g, and two 5 g masses.

Ordering information: This kit includes 4 sets of slotted masses, four 50 g mass hangers, and 4 table-clamp pulleys. A replacement force table pulley (470230-108) is available.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty