Would you like to log in again?
Ward’s® Organic Molecular Model Student Set, 134 Pieces
Supplier: VWR International
SET00603
470014-408EA 51.75 USD InStock
470014-408
Ward’s® Organic Molecular Model Student Set, 134 Pieces
Models Molecular Biology Models
Useful for individual or group study of molecules, these model sets offer an interactive learning opportunity.
  • Models Useful for Individual or Group Study
  • Interactive Learning Opportunity
  • Cost-Effective
  • Tool for Easy Assembly Included
  • Fifty Colorful Atom Parts and 64 Links Included
These model sets offer an interactive learning opportunity. Students construct molecules from color-coded plastic atoms and learn about the structure and function of each. The holes in each atom are set at angles that accept the included, flexible connectors of various lengths that represent the different bonds. Medium, gray links represent single covalent bonds, long and gray links represent double and triple covalent bonds, and short and white links can be used to construct compact models. With this comprehensive large set, students learn about alkanes, alcohols, aldehydes, ketones, and other organic structures.
This set features 50 colorful atom parts and 64 links. A tool for easy assembly is also included.
Ordering information: This set includes 12 carbon black 22 mm atoms, 20 hydrogen white 15 mm atoms, 6 oxygen red 20 mm atoms, 4 nitrogen blue 22 mm atoms, 1 sulfur yellow 22 mm atoms, 4 halogen green 20 mm atoms, 1 phosphorus purple 22 mm atoms, 1 metal gray 15 mm atoms, 52 medium links, 12 long and flexible links, and 1 tool. Measurements are in diameters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty