Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Spectrum Tubes

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PH1196B-7 SPTBR01 PH1196B9 STB-MX PH1196B-11 3376-16 PH1196B-8 3376-09 PH1196B-AIR PH1196B-6 PH1196B-1 SPTCL01 PH1196B-3 PH1196B-5 PH1196B PH1196B-4 PH1196B-2 PH1196B-10
470061-706EA 53.3 USD InStock
470061-706 470014-702 470230-432 470013-006 470164-816 470014-480 470014-482 470223-658 470014-470 470014-484 470014-472 470014-486 470014-474 470014-476 470005-836 470163-922 470005-834 470014-478
Spectrum Tubes
Educational Materials Physics Educational Materials Optics Learning Activities
Spectrum Tubes demonstrate the wave properties of different gases.

  • Ideal for Wavelength Determination
  • For Introducing Students to Better-Known Gases
  • Glass Tube Length: 10"
  • Capillary Portion Length: 4"
  • Tubes Work in a Variety of Spectroscopic Applications

Ideal for wavelength determination and introducing students to the spectra of better-known gases, these glass tubes are 10" long, and the capillary portion is about 4" long. The spectrum tube assemblies can be used in a variety of spectroscopic applications.

When they are observed through a spectroscope, the emission lines corresponding to the energy transition levels of the gaseous atoms or molecules appear as distinct components. The Spectrum Tube Power Supply is required but not included.

Ordering information: The mercury spectrum tube contains mercury and may not be available for sale in your state. Contact customer service for details.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty