Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal2 Microscopes, HM Series
Supplier: VWR
Boreal2
SK101 SK102 SK110
470014-514EA 463.6 USD InStock
470014-514 470016-038 470014-512
Boreal2 Microscopes, HM Series
Microscopes
With all-metal construction, a rugged, student-proof design, and precision optics, Boreal2 HM Microscopes are ideal for middle and high school students.
  • Rotatable, 45°-Inclined Viewing Head
  • Parfocal and Parcentered Optics
  • Lifetime Manufacturer's Warranty
  • Designed for Middle and High School Students
  • Total Magnification of 400x
A bit more advanced than the EM series, this microscope is designed for middle and high school students. Features include a gliding stage with one-touch stage clips, recessed eyepiece screws to prevent the disassembly of the microscope, a removable cord holder for easy storage, tension control to halt stage drift, large and easy-to-read magnification labels, and a reversed nose piece to protect objectives.
This microscope also features both a coarse and fine focus, has bright, white LED lighting, and offers a total magnification of 400x.
Ordering information: This product includes a manufacturer's warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty