Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal2 Microscopes, HM Advanced Series
Supplier: VWR
Boreal2
SK122 SK121 SK132 SK123BINOA
470014-518EA 559.5 USD InStock
470014-518 470014-528 470018-032 470014-520
Boreal2 Microscopes, HM Advanced Series
Microscopes
Perform quantitative experiments over a wide frequency range with these microscopes.
  • Objectives: 4x, 10x, 40x, and 100x
  • Large Magnification Labels
  • Protection against Disassembly
  • Tension Control
  • Charger Included
Bright, white, cold LED lighting and coated-glass optics give excellent images. The 4x, 10x, 40x, and 100x objectives have a wide color band with large, easy-to-read magnification labels on two sides for simplified viewing. Full-size, stable bodies hold a host of student-proof features, including locked-on stage clips, eye pieces, and objectives that offer protection against disassembly.
A four-hole reversed nosepiece protects objectives, and tension control stops stage drift. The microscopes are available in corded or cordless models; a removable cord holder is included on corded models for easy storage. The cordless model can be used anywhere for up to 50 hours with an eight-hour charge; a charger is included.
Ordering information: This series includes a lifetime warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty