Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal2 Advanced Stereomicroscopes
Supplier: VWR
Boreal2
SKS-30
470014-538EA 687.4 USD InStock
470014-538
Boreal2 Advanced Stereomicroscopes
Microscopes
New ergonomic design provides comfortable viewing and easy focus.
  • Inclined Binocular Head
  • Paired 10x Widefield Eyepieces with Diopter Adjustment in One
  • Built-in LED Illumination
  • Boreal Manufacturer's Lifetime Warranty
Conduct microscopy without shadows with this revolutionary LED incident light system for complete and even illumination of specimens with Boreal2 Advanced Stereomicroscopes. The three-way illumination features separate brightness control and superior optics. The zoom model also provides excellent images of demanding specimens.
Each stereomicroscope features adjustable interpupillary distance, one-touch stage clips, and a solid focus arm for ease of use. A heavy, stable base provides stable images, and the illuminator is protected by a sealed stage. A reversible head and carry handle offer easy, convenient storage.
Ordering information: This system comes in 110V or portable cordless versions. Corded models come with a removable cord rack.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty