Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal2 Digital Compound Microscopes - HM Series
Supplier: VWR
Boreal2
DM1000 SK1 DM-SK102
470014-546EA 1545.35 USD InStock
470014-546 470014-544
Boreal2 Digital Compound Microscopes - HM Series
Microscopes
These microscopes have all the advantages of the Boreal2 microscopes plus digital live image display, capture and manipulation.
  • Compatible with Windows 7 and above and Mac OSX and above
  • Motic image editing software included
  • One-year warranty on camera
  • Lifetime warranty on microscope chassis
These digital microscopes are both PC and Mac compatible, and the USB connection makes set up easy. Users can capture video or still images and even measure images on their screen. An auto-capture feature lets users collect images at pre-set intervals over time, which is perfect for prolonged class experiments.
Users can also export their results or save them for editing and annotating later. The exported images are compatible with other programs. Three different microscope models with different levels of resolution are available.
Ordering information: This product includes a manufacturer's warranty.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty