Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal Standard Compound Microscopes
Supplier: VWR
Boreal
MSH008 5584135 SFC-111LD 5745000 5745200 SFC-111LED 5584123
470145-716EA 367.7 USD InStock
470145-716 470177-090 470177-148 470145-560 470135-366 470145-714 470014-588
Boreal Standard Compound Microscopes
Microscopes
Boreal standard microscopes feature metal construction, a rugged student-proof design, and precision optics, making them a favorite among educators.
  • Coated-glass optics
  • Wide-field eyepiece with pointer
  • Lifetime warranty
This Boreal microscope's coated glass optics are parfocal and parcentered for a crisp, consistent field of view. Strategically located controls, a brass-on-steel focus guide, course and fine focus, a 360° rotatable head for viewing from any angle, a graduated mechanical stage, and an eyepiece pointer are all standard. Made of sturdy metal, it's designed to withstand years of wear and tear.
A reinforced joint between the head and eye tube prevents stripping, and a recessed eyepiece set screw prevents tampering. A slip clutch protects gears, and an adjustable, recessed stage stop protects optics and slides. Microscopes are 14¹/₂" tall.
Ordering information: This microscope includes a manual, a dust cover, and a manufacturer's warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty