Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Ward's® Introduction to Electrophoresis Lab System Set
Supplier: ALDON CORP SE
WARD470190-638 WARD470014-652 AA0089SET6
470101-112KT 17.4 USD InStock
470101-112 470014-652 470190-638
Ward's® Introduction to Electrophoresis Lab System Set
Educational Classroom Kits and Activities
This inexpensive set of materials outfits your whole class for gel electrophoresis at a great price.
  • Precursor to working with DNA and proteins
  • Material for six lab groups
  • Engaging activity for students
  • Estimated lab time: 45 minutes
  • Inexpensive system set
These electrophoresis dyes can be used as a precursor to working with DNA and proteins. When the colorful dyes are electrophoresed, they separate by their molecular size and charge. Students compare the electrophoresed results of the individual dyes to the dye mixture to find the missing component.
Demo Size requires 5 alkaline or rechargeable 9v batteries
Class Size requires 30 alkaline or rechargeable 9v batteries
Ordering information: This kit includes 6 electrophoresis boxes (class size) or 1 electrophoresis box (demonstration size), capillary pipettes, 6 different dyes, powdered agarose, TBE buffer, microfuge tubes, and instructions for students and teachers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty