Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boreal2 Stereomicroscope - EM Series
Supplier: VWR
1X SKS LED SKS-20
470014-656EA 320.79 USD InStock
470014-656 470039-602
Boreal2 Stereomicroscope - EM Series
Microscopes
Ideal for younger students
  • 2X objectives
  • Optional 1X, 3X, and 4X objectives available
  • Boreal Lifetime Warranty
This stereomicroscope offers a long range of focus for large and small samples. An 80 mm working distance is offered at 20X magnification for versatility in specimen studies. To protect the microscope, black, white, and translucent stage plates fit on the sealed stage to keep liquids out of the base. The ergonomic design includes a carry handle for easy transport and a large metal body with stable base for sturdy durability.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty