Would you like to log in again?
Basic Van de Graaff Generator (Accessories Only)
Supplier: SCIENCE INTERACTIVE GROUP
615-3110
470014-736EA 20.8 USD InStock
470014-736
Basic Van de Graaff Generator (Accessories Only)
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Create Dramatic Electrostatic Effects
Please Note: Only Basic Van de Graaff Accessories available.
Students always enjoy creating sparks and fanning out a classmate’s hair with static from a Van de Graaff generator. Now you can generate up to 200,000 volts and five-inch sparks, even in 90% humidity, easily and inexpensively. The economical generator has a plastic molded base and housing that lifts off to reveal the working parts; the neoprene belt lasts up to 500 hours. The set includes an aluminum globe, PVC column, ground terminal, neoprene belt, and 16 page manual. 110 VAC . Some assembly is required. Size: 8"L x 8"W x 12"H.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty