Would you like to log in again?
Pocket Hardness Points Set
Supplier: VWR
WARD470014-828
470014-828KT 107.25 USD InStock
470014-828
Pocket Hardness Points Set
Specimens Rock Specimens Specialty Rocks
Students determine a mineral's hardness between five and 10 on Mohs' scale.
  • Students Determine a Mineral's Hardness
  • Seven Cut Minerals Mounted on Four Rods
  • Rods Stamp with Each Mineral's Hardness
The neat little case tucks into a pocket, so students have an accurate gauge of harder minerals ready. Seven cut minerals, representing Mohs’ hardness measures of 5, 6, 7, 8, 8 1/2, 9, and 10, are permanently mounted on the ends of four pen-size aluminum rods.
The rods are stamped with each mineral’s hardness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty