Would you like to log in again?
Ward's® Single Probes Wi-Fi Module
Supplier: EISCO SCIENTIFIC LLC
WSP-WIFI-500
470014-874EA 167.35 USD InStock
470014-874
Ward's® Single Probes Wi-Fi Module
Temperature Probes
Wirelessly transmit data from your Ward's Single Probe Base Unit and Sensor to any Wi-Fi-enabled device, including PC/Mac, iPad, Android tablet, or Smartphone.
  • Wireless capability
  • Fully-functional hotspot
  • Transmit data to up to 5 devices
Simply plug in to the base unit to broadcast a self-contained wireless network compatible with any Wi-Fi-enabled device in your classroom. Also a fully-functional hotspot, this module requires no Internet connection or external network for use. One module will transmit data simultaneously to up to five devices and device types that are connected to the self-contained module network so students can see the same data set throughout the entire classroom; to transmit multiple date sets to multiple devices in your classroom, simply add another Wi-Fi Module to another Ward's Single Probes Base Unit and Sensor combo.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty