Would you like to log in again?
Spray Chalk
CKS-RD
470014-898EA 27.29 USD InStock
470014-898
Spray Chalk
Educational Materials Forensics Educational Materials Forensics Supplies
Ideal Crime Scene Marking Solution
Similar in composition to regular chalk, this non-toxic Spray Chalk produces a bright, clearly visible, fluorescent mark. It will adhere to nearly any surface, including asphalt, concrete, brick, wood, grass and metal; but it is not permanent, so it can be easily removed. Size: 7 oz.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty