Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ward's® Instant Water Quality Test Kit
Supplier: Avantor
WARD470014-998
470014-998EA 45.99 USD InStock
470014-998
Ward's® Instant Water Quality Test Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Test Five Factors on One Test Strip
Your students will be amazed to learn that their drinking glasses may hold more than just water. Have them bring water samples to class or take the test into the field, and this water quality testing kit will do the rest. Step-by-step instructions, as well as background information on each testing parameter is included. Test for total and free chlorine, total hardness, total alkalinity, and pH simultaneously on one test strip. The kit includes 50 test strips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty