Would you like to log in again?
Ward's® Gravity and Orbits Lab Activity
Supplier: Avantor
WARD470015-222
470015-222EA 122.59 USD InStock
470015-222
Ward's® Gravity and Orbits Lab Activity
Educational Classroom Kits and Activities
Enough Materials to Construct Five Sets of Two Models
  • Includes a teacher's guide
Students investigate the effect of gravity on a moving body, then determine the relationship between a celestial object’s period of revolution and the force pulling on the object, by constructing two simple models in a two-part lab activity. You will receive enough materials for five setups, a teacher’s guide, and student copymasters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty