Would you like to log in again?
Ward's® The Road to Life: Forming Coacervates Lab Activity
Supplier: Avantor
WARD470015-316 WARD470007-678
470007-678KT 83.99 USD InStock
470007-678 470015-316
Ward's® The Road to Life: Forming Coacervates Lab Activity
Educational Classroom Kits and Activities
Provide insight into evolution.
  • Students will study the formation of coacervates
In this exceptional lab activity, your class actually produces coacervates, which are liquid colloidal suspensions of organic compounds. Coacervates, while not truly alive, exhibit several lifelike properties. They contain an outer membrane and grow by reacting to the physical and chemical conditions of their environment. Students will study the formation of coacervates, stain them and compare them to simple living organisms. The kit contains materials for 30 setups, a teacher’s guide, and student copy master.
Ordering information: A refill kit is available and contains only consumable component items. It does not contain additional student or teacher guides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty