Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ward's® Plasmolysis in Plant Cells Lab Activity
Supplier: Avantor
WARD470015-332
470015-332KT 73.69 USD InStock
470015-332 470328-416
Ward's® Plasmolysis in Plant Cells Lab Activity
Educational Classroom Kits and Activities
Observe Dramatic Effect of Water Loss on Onion Cells.
  • Aligned to NGSS standards (MS/HS: LS1)
What makes a plant wilt when it needs water, and “come back to life” when it receives that water? Find out with this simple investigation. Using only an onion you supply and this kit’s materials, students can easily see the effect of varying osmotic conditions on plant cells, and learn the importance of water to a plant’s structure. It includes enough materials for six setups, a teacher’s guide, and student copymaster.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty