Would you like to log in again?
Ward's® Do Microbes Arise Spontaneously? Lab Activity
Supplier: Avantor
WARD470015-336
470015-336KT 81.19 USD InStock
470015-336
Ward's® Do Microbes Arise Spontaneously? Lab Activity
Educational Classroom Kits and Activities
Determine the Origins of Microbes by Re-creating a Classic Experiment
Scientists once believed that creatures developed from a “vital force” in the air. This concept of spontaneous generation was accepted for centuries until, in the 1800s, Louis Pasteur proved that life could be created only from other life. Now your students can re-create Pasteur’s experiment as they observe two samples of nutrient broth, one sterile and the other nonsterile, to find out where the bacteria grows. The activity comes with materials for six setups, teacher’s guide, and student copymaster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty