Would you like to log in again?
Ward's® Introductory Tissue Culture Lab Activity
Supplier: Avantor
470015-382KT 97.09 USD InStock
470015-382
Ward's® Introductory Tissue Culture Lab Activity
Educational Classroom Kits and Activities
Study Animal Cells in Detail
  • Detailed procedure for chick cell isolation
  • Develop aseptic technique
  • Materials for 8 lab groups
  • Time Required: 120 minutes over 1-2 weeks
Using methods to initiate primary cell cultures from fresh embryonic tissue, students acquaint themselves with growing cells in vitro, handling tissue cultures, and observing patterns of cell growth, rates of growth, general appearance, and distinguishing characteristics.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Required but not included: fresh fertilized eggs from local farm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty