Would you like to log in again?
Ward's® Bread Mold Culture and Study Kit
Supplier: Avantor
470015-774KT 79.89 USD InStock
470015-774
Ward's® Bread Mold Culture and Study Kit
Educational Classroom Kits and Activities
Culture and identify different types of mold
  • Culture molds from bread and compare them to pure samples we send you
  • Introduce the concept of the dichotomous key
  • No need to cook or dispense media
  • This activity can be done without having mastered sterile technique.
Beginning with the initial culture, grown on slices of bread, students observe the development of mold, then use the included dichotomous key to identify the types of mold that appear, as well as compare their growth rates and morphological characteristics.
The kit includes demonstration plate cultures of Aspergillus niger, Mucor heimalis, Penicillium notatum, and Rhizopus stolonifer; teacher’s guide; and student copymaster with dichotomous key and additional suggested activities.
Includes: coupon for perishable materials.
Ordering information: This item contains a copupon for live or perishable materials. You will need to provide a date that the live or perishable components will arrive.
Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground.
Perishable components will ship via overnight or second day delivery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty