Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Primary and Secondary Coil

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PH50645
470015-940EA 84.55 USD InStock
470015-940
Primary and Secondary Coil
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Primary and Secondary Coils are ideal for student experiments on transformer effects.

  • Soft Iron Core Provided
  • Binding Posts for Both Coils Provided
  • Inner Coil Inductance: 78 ± 22µH
  • Inner Coil Resistance: 0.4 ± 0.1Ω
  • Outer Coil Inductance: 63 ±3mH
  • Outer Coil Resistance: 97 ±2Ω

The inner coil has heavier wire and serves as the primary coil. It has 150 turns, its inductance is 78 ± 22 µH, has a resistance of 0.4 ± 0.1Ω, and its capacitance is 142±2pF. Its base is 4.5 x 4.5 cm.

The outer coil has fine, insulated wire on a plastic spool with square ends to support it vertically or horizontally. It has 3300 turns, its inductance is 63 ±3 mH, its resistance is 97 ±2Ω, and its capacitance is 124 ±2pF. Its base is 4.5 x 4.5 cm.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty