Would you like to log in again?
Resistance Substitution Apparatus
Supplier: ELENCO ELECTRONICS INC SE
RS-500
470016-290EA 150.5 USD InStock
470016-290
Resistance Substitution Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Resistance Substitution Apparatus is convenient for general-purpose resistance work.
  • Watt Rating: 1W
  • Resistance Range: 1 to 11,000,000 ohms
  • Accuracy: ±1%
  • Twenty-Eight Slide Switches
  • Three Five-Way Binding Posts
This compact resistance box features 28 slide switches on the front, so users can set the resistance in 1-ohm steps. The resistors have a 1% tolerance. The Apparatus can reach a range of one to over 11 million ohms. Three five-way binding posts are included, one of which grounds the case.
It has a watt rating of 1W, a resistance range of 1 to 11,000,000 ohm, and an accuracy rate of±1%. Its dimensions are 10.5 x 16 x 3.5 cm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty