Would you like to log in again?
Fundamentals of Physics Pendulum Kit
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PND08
470016-330EA 56.5 USD InStock
470016-330
Fundamentals of Physics Pendulum Kit
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
This Pendulum kit is an excellent model for investigating energy.
  • Large-Scale Protractor
  • Easy-To-Change Masses Included
  • Teacher's Manual Included
  • Adjustable String Length
  • Students Learn How Different Factors Affect a Pendulum's Period
Users can adjust string length and the mass of the pendulum. A large protractor makes angle measurements easy to read, and the photogate system can be attached at the bottom of the swing arc to record the period.
Students can investigate how the angle of release, mass, and string length affect the period of a pendulum.
Ordering information: This kit includes a protractor backboard, string, pendulum, mounting screw, and activity guide. The Fun Stand and Photogate System are required but not provided.
Caution: Caution:Small parts can cause a choking hazard. This product is not for children under three years.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty