Would you like to log in again?
Uniform Circular Motion Demonstration
Supplier: Avantor
WARD470016-810
470016-810EA 128.99 USD InStock
470016-810
Uniform Circular Motion Demonstration
Educational Classroom Kits and Activities
Give Your Circular Motion Lessons a Lift!
  • Easy to use
  • Includes instructions
Developed in partnership with educator Jeffrey Wetherhold, this motorized toy airplane helps exhibit properties of tension, weight, speed, and circular motion in this engaging lab. Tethered to the ceiling with two C clamps, the airplane, attached to by a string, moves in uniform horizontal turns. Combining ideals of physics and math, students use Newton's Second Law of Motion in addition to algebra and trigonometry to derive their own equation for predicting the tension of the string. 
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty