Would you like to log in again?
Ward's® Chemistry Rainbow Down the Tube Demonstration
Supplier: Avantor
WARD470016-836
470016-836EA 49.99 USD InStock
470016-836
Ward's® Chemistry Rainbow Down the Tube Demonstration
Educational Classroom Kits and Activities
Somewhere Over the Rainbow Chemistry Lessons Fly.
  • Instruction manual included
  • Teach density, solubility, and pH with one demonstration!
Engage your students with this imaginative activity that allows you to pour a beautiful rainbow down the inside of your Ward's® Chemistry Demonstration tube. A concentrated solution of sodium carbonate will be added to a dilute solution of hydrochloric acid and universal indicator. You can then teach density, solubility, migration of ions, and pH as the dense solution travels down the tube. A glass demonstration tube is required and sold separately.
Ordering information: Includes Sodium Carbonate, Hydrochloric Acid, Universal Indicator, and Instructions. Required but Not Provided are Glass Demonstration Tube, and General Glassware.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty