Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Ward's® Chemistry Instant Light in the Bag Inquiry Activity
Supplier: Avantor
LL0255-100G WARD470016-838
470016-838EA 40.99 USD InStock
470016-838 470301-302
Ward's® Chemistry Instant Light in the Bag Inquiry Activity
Educational Classroom Kits and Activities
With this activity, teachers just add water to a dry powder for an intriguing, instant illumination in a bag.
  • Demonstrate chemiluminescence
  • Quick, easy, mess-free demonstration
Just add water to a dry powder for an intriguing instant illumination in a bag. The mixture glows in the dark and allows students to discover the chemistry behind chemiluminescence and explore it by designing experiments to make the light brighter or last longer. Students will begin to understand that the light is a product and the kinetics of the reaction can be controlled by temperature. A great introduction to kinetics, scientific inquiry, and chemiluminescence. Includes 100 g bottle of instant light crystals, instructions and 10 heavy duty plastic bags.
Students begin to understand that the light is a product, and the kinetics of the reaction can be controlled by temperature. The activity is a great introduction to kinetics, scientific inquiry, and chemiluminescence.
Ordering information: This kit includes 100 g bottle of instant light crystals, instructions, and 10 heavy duty plastic bags.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty