Would you like to log in again?
Vernier EasyLink® Interface
Supplier: VERNIER SOFTWARE & TECH SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Vernier
EZ-LINK
470017-138EA 78.99 USD InStock
470017-138
Vernier EasyLink® Interface
Electrodes Other Electrodes
Inexpensive data collection direct to any TI-Nspire® handheld or TI-84 Plus graphing calculator.
  • Connects BTA (sensor) to mini-A USB (calculator)
  • Compatible with all models of TI-84 Plus, TI-84 Plus CE, and TI-Nspire® CXII Texas Instruments calculators
  • Supports over 62 analog Vernier sensors (Digital sensors and microphone are incompatible)
EasyLink® is a single-channel interface that plugs into the USB port of a TI-84 Plus graphing Calculator or TI-Nspire® Handheld. Use EasyLink® and a supported sensor to explore math and science concepts found in real-world phenomena such as light, pressure, force, and more.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty