Would you like to log in again?
Vernier® LabQuest Mini®
Supplier: VERNIER SOFTWARE & TECH SE
LQ-MINI
470017-584EA 188.99 USD InStock
470017-584
Vernier® LabQuest Mini®
Data Loggers
LabQuest Mini is powerful, affordable, and easy to use. It brings the power of Vernier's award-winning LabQuest technology to teachers who don't need the versatility of a standalone device. It is the perfect solution for educators collecting data with a windows or macintosh computer.
  • Maximum sampling rate: 100 kHz
  • Five sensor ports
  • Over 70 compatible sensors
  • Analog inputs: 3
  • Digital inputs: 2
  • Logger Pro 3, logger lite, and graphical analysis for chrome compatible
LabQuest Mini software has unparalleled power, analysis, and curricular support. It is for educational use only. USB used for computer connection, full speed of 2.0.
Maximum sampling rate is 100000 samples per second. Power is supplied by the USB port of the computer - auxiliary AC power or batteries are not required.
Delivery information: It includes computer connection (USB) cable and user manual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty