Would you like to log in again?
Science Take-Out® Mark-Recapture Population Sampling
Supplier: Science Take-Out
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
111-NOTG 111 111U
470017-740EA 13.7 USD InStock
470017-740 470017-780 470017-741
Science Take-Out® Mark-Recapture Population Sampling
Educational Materials Biology Educational Materials Ecology Learning Activities
How Many Spiders Are Hiding In The Leaf Litter?
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
How many plastic spiders are hiding in simulated “leaf litter”? Use the mark-recapture population sampling technique to estimate the number of organisms in a population. Capture, mark and release a random sample of spider in your "leaf litter". Recapture a second random sample of spiders and use a mathematical equation to estimate the total number of animals in the population. This complete "dry lab" activity contains all required materials.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty