Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Science Take-Out® Yeast Populations
Supplier: Science Take-Out
123R 123-NOTG 123U 123
470017-804PK 103.55 USD InStock
470017-804 470017-764 470017-864 470017-765
Science Take-Out® Yeast Populations
Educational Materials Biology Educational Materials Microbiology Learning Activities
What Environmental Conditions Can Affect Organism Growth?
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
Test samples from a simulated yeast population to determine yeast population size, the pH of the environment, and the amount of sugar present. Graph your results and compare them with results from another researcher. This complete experiment setup contains all materials and does not require lab equipment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty