Would you like to log in again?
Science Take-Out® Kidney Dialysis
Supplier: Science Take-Out
101-NOTG 101 101R 101U
470017-806EA 54.8 USD InStock
470017-806 470017-868 470017-869 470017-766
Science Take-Out® Kidney Dialysis
Educational Materials Biology Educational Materials Physiology Learning Activities
Create A Model Of A Kidney Dialysis Machine.
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
How does a kidney dialysis machine remove wastes from the blood of a patient with kidney failure? Create a mini-model of a dialysis machine. Determine what substances diffuse from the patient into the machine. Determine what substances should be added to the fluid in the dialysis machine to prevent the loss of essential materials. Compare hemodialysis (using a dialysis machine) to peritoneal dialysis (an alternative treatment for kidney failure). This complete experiment setup contains all materials and does not require lab equipment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty