Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier Charge Sensor

Supplier: VERNIER SOFTWARE & TECH SE
Vernier
CRG-BTA
470017-894EA 88.99 USD InStock
470017-894
Vernier Charge Sensor
Electrodes Other Electrodes
Electronic electroscope to obtain quantitative measurements of charge.

  • Wired connection with BTA plug
  • Maximum input ±150 V
  • Typical bias input current 0.005 pA
  • Input capacitance 0.01 μF
  • Instrument time constant 0.1 s

Unlike a traditional electroscope, the Charge Sensor can make quantitative measurements. Numerical measurements improve many electrostatics experiments, such as charging by induction, charging by friction, and charging by contact.

The sensor can also be used to measure charge polarity. An extremely high-impedance voltage sensor with a 0.01 μF input capacitor makes these measurements possible. The sensor has two operating ranges and a zeroing switch to discharge the input capacitor.

Ordering information: LabQuest® (2, 3, Mini) interface is required but not included.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty