Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Physiology with Vernier Lab Book
Supplier: VERNIER SOFTWARE & TECH SE
HP-A
470018-202EA 51.99 USD InStock
470018-202
Human Physiology with Vernier Lab Book
Charts
Step-by-Step Experiments That Are Easily Executable
Vernier makes teaching Human physiology easy by providing 24 experiments that can be implemented in conjunction with your Vernier probeware. Students will be more acquainted with their bodies as they learn how all the organ systems work together by measuring the activity of the heart, lungs, nervous system, musculoskeletal system and the upper respiratory tract. Homeostatis is also a prominent theme, with many of the included experiments addressing how the body tries to maintain its equilibrium while reacting in response to stress, sickness, or other external influences. Word processing files of the student sections can be found on the included CD.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty