Would you like to log in again?
Fiber Optic Holder for Carousel Power Supply
Supplier: STEINER ENTERPRISES SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SE1041-00
470018-232EA 21.7 USD InStock
470018-232
Fiber Optic Holder for Carousel Power Supply
Educational Materials Physics Educational Materials Optics Learning Activities
Dual purpose for Dual Benefits
  • The Fiber Optics Holder is an accessory for the Carousel Spectrum Tube Power Supply with two great features
The Fiber Optics Holder holds the end of the fiber optic probe of a digital spectrometer in place when in front of the illuminated spectrum tube in the Carousel Power Supply, allowing the probe to be positioned optimally to receive the maximum amount of light from the tube. It also holds the probe end at a consistent safe distance from the tube surface.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty