Would you like to log in again?
Magdeburg Hemispheres
Supplier: VWR International
PH0171A 6132-0010
470211-868CS 1096.95 USD InStock
470211-868 470018-246
Magdeburg Hemispheres
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Employs Guericke’s Method for Demonstrating the Force of Atmospheric Pressure.
  • Two plastic, 10cm-diameter hemispheres, each with a sturdy handle, make up this device
  • Operating instructions are included
One hemisphere has a gasket seal; the other has a stopcock for attaching to 0.8-1.2cm inner-diameter vacuum tubing. Join the hemispheres, hook them up to a hand pump, aspirator, or vacuum pump, and evacuate them. Students will find it hard, if not impossible, to pull them apart. Open the stopcock to let air in, and the hemispheres separate easily.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty