Would you like to log in again?
Low Frequency Generator
Supplier: VWR International
PH1298-N9
470018-260EA 323.1 USD InStock
470018-260
Low Frequency Generator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Reliable and User Friendly
  • Frequency range : 1HZ-2 kHz.
  • Waveform: sine square and triangular. TTL output: controllable ±13 Volts when auxiliary input of 15 Volts is used, 9 volts if batteries are used.
  • Can use an external power supply or two 9V batteries, not included.
Two potentiometers for frequency and amplitude are featured on this generator along with two dials for selection of frequency and waveform. Two outputs - the alternating signal and the TTL signal are also included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty